Merck
  • Selective Ru(II)/lawsone complexes inhibiting tumor cell growth by apoptosis.
Selective Ru(II)/lawsone complexes inhibiting tumor cell growth by apoptosis.
Journal of inorganic biochemistry (2017-09-04)
Katia M Oliveira, Luna-Dulcey Liany, Rodrigo S Corrêa, Victor M Deflon, Marcia R Cominetti, Alzir A Batista
ABSTRACT
New Ru(II) complexes with lawsone (law) characterized as trans-[Ru(law)(PPh
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
2-Hydroxy-1,4-naphthoquinone, 97%
Sigma-Aldrich
4-4′-Dimethoxy-2-2′-bipyridine, 97%