Merck
The Neurobeachin-like 2 Protein Regulates Mast Cell Homeostasis.
Journal of immunology (Baltimore, Md. : 1950) (2017-09-10)
Sebastian Drube, Randy Grimlowski, Carsten Deppermann, Julia Fröbel, Florian Kraft, Nico Andreas, David Stegner, Jan Dudeck, Franziska Weber, Mandy Rödiger, Christiane Göpfert, Julia Drube, Daniela Reich, Bernhard Nieswandt, Anne Dudeck, Thomas Kamradt
ABSTRACT
The neurobeachin-like 2 protein (Nbeal2) belongs to the family of beige and Chediak-Higashi (BEACH) domain proteins. Loss-of-function mutations in the human
Sigma-Aldrich
Anti -Dinitrophenyl antibody, Mouse monoclonal, IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Albumin, dinitrophenyl, lyophilized powder