Merck
Boeravinone B, A Novel Dual Inhibitor of NorA Bacterial Efflux Pump of
Frontiers in microbiology (2017-10-20)
Samsher Singh, Nitin P Kalia, Prashant Joshi, Ajay Kumar, Parduman R Sharma, Ashok Kumar, Sandip B Bharate, Inshad A Khan
ABSTRACT
This study elucidated the role of boeravinone B, a NorA multidrug efflux pump inhibitor, in biofilm inhibition. The effects of boeravinone B plus ciprofloxacin, a NorA substrate, were evaluated in NorA-overexpressing, wild-type, and knocked-out
Sigma-Aldrich
2-Phenylindole, technical grade, 95%