Merck
  • Sterol regulatory element-binding protein 1 inhibitors decrease pancreatic cancer cell viability and proliferation.
Sterol regulatory element-binding protein 1 inhibitors decrease pancreatic cancer cell viability and proliferation.
Biochemical and biophysical research communications (2017-05-10)
Siqingaowa, Sathiya Sekar, Venkat Gopalakrishnan, Changiz Taghibiglou
ABSTRACT
Sterol regulatory element-binding protein1 (SREBP1) is a key regulatory factor that controls lipid homeostasis. Overactivation of SREBP1 and elevated lipid biogenesis are considered the major characteristics in malignancies of prostate cancer, endometrial cancer, and glioblastoma. However, the impact of SREBP1 activation in the progression of pancreatic cancer has not been explored. The present study examines the effect of suppression of SREBP1 activation by its inhibitors like fatostatin and PF429242 besides analyzing the impact of inhibitory effects on SREBP1 downstream signaling cascade such as fatty acid synthase (FAS), hydroxymethylglutaryl-CoA reductase (HMGCoAR), stearoyl-CoA desaturase-1 (SCD-1), and tumor suppressor protein p53 in MIA PaCa-2 pancreatic cancer cells. Both fatostatin and PF429242 inhibited the growth of MIA PaCa-2 cells in a time and concentration-dependent manner with maximal inhibition attained at 72 h time period with IC
Sigma-Aldrich
PF-429242 dihydrochloride, ≥98% (HPLC)