Merck
  • Structural basis for functional selectivity and ligand recognition revealed by crystal structures of human secreted phospholipase A
Structural basis for functional selectivity and ligand recognition revealed by crystal structures of human secreted phospholipase A
Scientific reports (2017-09-09)
Shulin Hou, Tingting Xu, Jinxin Xu, Linbing Qu, Yong Xu, Ling Chen, Jinsong Liu
ABSTRACT
Secreted phospholipases A