Merck
  • Influence of Lattice Polarizability on the Ionic Conductivity in the Lithium Superionic Argyrodites Li
Influence of Lattice Polarizability on the Ionic Conductivity in the Lithium Superionic Argyrodites Li
Journal of the American Chemical Society (2017-07-26)
Marvin A Kraft, Sean P Culver, Mario Calderon, Felix Böcher, Thorben Krauskopf, Anatoliy Senyshyn, Christian Dietrich, Alexandra Zevalkink, Jürgen Janek, Wolfgang G Zeier
ABSTRACT
In the search for novel solid electrolytes for solid-state batteries, thiophosphate ionic conductors have been in recent focus owing to their high ionic conductivities, which are believed to stem from a softer, more polarizable anion framework. Inspired by the oft-cited connection between a soft anion lattice and ionic transport, this work aims to provide evidence on how changing the polarizability of the anion sublattice in one structure affects ionic transport. Here, we systematically alter the anion framework polarizability of the superionic argyrodites Li
Sigma-Aldrich
Lithium sulfide, 99.98% trace metals basis