Merck
  • High-Throughput Screens to Discover Small-Molecule Modulators of Ryanodine Receptor Calcium Release Channels.
High-Throughput Screens to Discover Small-Molecule Modulators of Ryanodine Receptor Calcium Release Channels.
SLAS discovery : advancing life sciences R & D (2016-10-21)
Robyn T Rebbeck, Maram M Essawy, Florentin R Nitu, Benjamin D Grant, Gregory D Gillispie, David D Thomas, Donald M Bers, Razvan L Cornea
ABSTRACT
Using time-resolved fluorescence resonance energy transfer (FRET), we have developed and validated the first high-throughput screening (HTS) method to discover compounds that modulate an intracellular Ca
Sigma-Aldrich
Diazoxide
Supelco
Cefixime trihydrate, suitable for microbiology, for ion-selective electrodes, ≥98.0% (HPLC)
Sigma-Aldrich
5,7-Dichloro-8-quinolinol, 99%