Merck
Rapid, label-free CD4 testing using a smartphone compatible device.
Lab on a chip (2017-07-14)
Manoj Kumar Kanakasabapathy, Hardik J Pandya, Mohamed Shehata Draz, Manjyot Kaur Chug, Magesh Sadasivam, Shreya Kumar, Behzad Etemad, Vinish Yogesh, Mohammadali Safavieh, Waseem Asghar, Jonathan Z Li, Athe M Tsibris, Daniel R Kuritzkes, Hadi Shafiee
ABSTRACT
The most recent guidelines have called for a significant shift towards viral load testing for HIV/AIDS management in developing countries; however point-of-care (POC) CD4 testing still remains an important component of disease staging in multiple developing countries. Advancements in micro/nanotechnologies and consumer electronics have paved the way for mobile healthcare technologies and the development of POC smartphone-based diagnostic assays for disease detection and treatment monitoring. Here, we report a simple, rapid (30 minutes) smartphone-based microfluidic chip for automated CD4 testing using a small volume (30 μL) of whole blood. The smartphone-based device includes an inexpensive (<$5) cell phone accessory and a functionalized disposable microfluidic device. We evaluated the performance of the device using spiked PBS samples and HIV-infected and uninfected whole blood, and compared the microfluidic chip results with the manual analysis and flow cytometry results. Through t-tests, Bland-Altman analyses, and regression tests, we have shown a good agreement between the smartphone-based test and the manual and FACS analysis for CD4 count. The presented technology could have a significant impact on HIV management in developing countries through providing a reliable and inexpensive POC CD4 testing.
Product Description
BRAND® counting chamber BLAUBRAND® Neubauer improved, without clips, double ruled
Sigma-Aldrich
Sodium acetate, anhydrous, ≥99%