- Erythritol is a pentose-phosphate pathway metabolite and associated with adiposity gain in young adults.
Erythritol is a pentose-phosphate pathway metabolite and associated with adiposity gain in young adults.
Proceedings of the National Academy of Sciences of the United States of America (2017-05-10)
Katie C Hootman, Jean-Pierre Trezzi, Lisa Kraemer, Lindsay S Burwell, Xiangyi Dong, Kristin A Guertin, Christian Jaeger, Patrick J Stover, Karsten Hiller, Patricia A Cassano
PMID28484010
ABSTRACT
Metabolomic markers associated with incident central adiposity gain were investigated in young adults. In a 9-mo prospective study of university freshmen (
Product Description
Supelco
Alkane standard mixture for performance tests of GC-systems, analytical standard, C10 - C40 (all even), 50 mg/l each