Merck
  • Purification and development of ELISAs for two forms of vitellogenin in Indian walking catfish, Clarias batrachus (L.).
Purification and development of ELISAs for two forms of vitellogenin in Indian walking catfish, Clarias batrachus (L.).
Fish physiology and biochemistry (2017-03-02)
S Mahapatra, Sk Kabita, D Bhattacharya, S Sarkar, S K Juin, S Maitra, P Nath
ABSTRACT
Two forms of vitellogenin (Vg: Vg1 and Vg2) were purified from the plasma of estradiol-17β (E