MilliporeSigma
  • Toxicogenomic responses of human alveolar epithelial cells to tungsten boride nanoparticles.
Toxicogenomic responses of human alveolar epithelial cells to tungsten boride nanoparticles.
Chemico-biological interactions (2017-07-02)
Hasan Türkez, Mehmet Enes Arslan, Erdal Sönmez, Abdulgani Tatar, Metin Açikyildiz, Fatime Geyikoğlu
ABSTRACT
During the recent years, microarray analysis of gene expression has become an inevitable tool for exploring toxicity of drugs and other chemicals on biological systems. Therefore, toxicogenomics is considered as a fruitful area for searching cellular pathways and mechanisms including cancer, immunological diseases, environmental responses, gene-gene interactions and chemical toxicity. In this work, we examined toxic effects of Tungsten Borides NPs on gene expression profiling of the human lung alveolar epithelial cells (HPAEpiC). In line with this purpose, a single crystal of tungsten boride (mixture of WB and W