Merck
  • Lecithin:Cholesterol Acyltransferase Activation by Sulfhydryl-Reactive Small Molecules: Role of Cysteine-31.
Lecithin:Cholesterol Acyltransferase Activation by Sulfhydryl-Reactive Small Molecules: Role of Cysteine-31.
The Journal of pharmacology and experimental therapeutics (2017-06-04)
Lita A Freeman, Stephen J Demosky, Monika Konaklieva, Rostislav Kuskovsky, Angel Aponte, Alice F Ossoli, Scott M Gordon, Ross F Koby, Kelly A Manthei, Min Shen, Boris L Vaisman, Robert D Shamburek, Ajit Jadhav, Laura Calabresi, Marjan Gucek, John J G Tesmer, Rodney L Levine, Alan T Remaley
ABSTRACT
Lecithin:cholesterol acyltransferase (LCAT) catalyzes plasma cholesteryl ester formation and is defective in familial lecithin:cholesterol acyltransferase deficiency (FLD), an autosomal recessive disorder characterized by low high-density lipoprotein, anemia, and renal disease. This study aimed to investigate the mechanism by which compound A [3-(5-(ethylthio)-1,3,4-thiadiazol-2-ylthio)pyrazine-2-carbonitrile], a small heterocyclic amine, activates LCAT. The effect of compound A on LCAT was tested in human plasma and with recombinant LCAT. Mass spectrometry and nuclear magnetic resonance were used to determine compound A adduct formation with LCAT. Molecular modeling was performed to gain insight into the effects of compound A on LCAT structure and activity. Compound A increased LCAT activity in a subset (three of nine) of LCAT mutations to levels comparable to FLD heterozygotes. The site-directed mutation LCAT-Cys31Gly prevented activation by compound A. Substitution of Cys31 with charged residues (Glu, Arg, and Lys) decreased LCAT activity, whereas bulky hydrophobic groups (Trp, Leu, Phe, and Met) increased activity up to 3-fold (
Sigma-Aldrich
LCAT activator compound A, ≥98% (HPLC)