Merck
MC4R-dependent suppression of appetite by bone-derived lipocalin 2.
Nature (2017-03-09)
Ioanna Mosialou, Steven Shikhel, Jian-Min Liu, Antonio Maurizi, Na Luo, Zhenyan He, Yiru Huang, Haihong Zong, Richard A Friedman, Jonathan Barasch, Patricia Lanzano, Liyong Deng, Rudolph L Leibel, Mishaela Rubin, Thomas Nickolas, Wendy Chung, Lori M Zeltser, Kevin W Williams, Jeffrey E Pessin, Stavroula Kousteni
ABSTRACT
Bone has recently emerged as a pleiotropic endocrine organ that secretes at least two hormones, FGF23 and osteocalcin, which regulate kidney function and glucose homeostasis, respectively. These findings have raised the question of whether other bone-derived hormones exist and what their potential functions are. Here we identify, through molecular and genetic analyses in mice, lipocalin 2 (LCN2) as an osteoblast-enriched, secreted protein. Loss- and gain-of-function experiments in mice demonstrate that osteoblast-derived LCN2 maintains glucose homeostasis by inducing insulin secretion and improves glucose tolerance and insulin sensitivity. In addition, osteoblast-derived LCN2 inhibits food intake. LCN2 crosses the blood-brain barrier, binds to the melanocortin 4 receptor (MC4R) in the paraventricular and ventromedial neurons of the hypothalamus and activates an MC4R-dependent anorexigenic (appetite-suppressing) pathway. These results identify LCN2 as a bone-derived hormone with metabolic regulatory effects, which suppresses appetite in a MC4R-dependent manner, and show that the control of appetite is an endocrine function of bone.
Sigma-Aldrich
MISSION® esiRNA, targeting human MC4R