Merck
  • Synthesis and Characterization of Diol-Based Unsaturated Polyesters: Poly(diol fumarate) and Poly(diol fumarate-co-succinate).
Synthesis and Characterization of Diol-Based Unsaturated Polyesters: Poly(diol fumarate) and Poly(diol fumarate-co-succinate).
Biomacromolecules (2017-05-11)
Alexander M Tatara, Emma Watson, Tejus Satish, David W Scott, Dimitrios P Kontoyiannis, Paul S Engel, Antonios G Mikos
ABSTRACT
In this work, we describe the synthesis and characterization of variants of poly(diol fumarate) and poly(diol fumarate-co-succinate). Through a Fischer esterification, α,ω-diols and dicarboxylic acids were polymerized to form aliphatic polyester comacromers. Because of the carbon-carbon double bond of fumaric acid, incorporating it into the macromer backbone structure resulted in unsaturated chains. By choosing α,ω-diols of different lengths (1,6-hexanediol, 1,8-octanediol, and 1,10-decanediol) and controlling the amount of fumaric acid in the dicarboxylic acid monomer feed (33, 50, and 100 mol %), nine diol-based macromer variants were synthesized and characterized for molecular weight, number of unsaturated bonds per chain, and thermal properties. Degradation and in vitro cytotoxicity were also measured in a subset of macromers. As proof-of-principle, macromer networks were photo-cross-linked to demonstrate the ability to perform free radical addition using the unsaturated macromer backbone. Cross-linked macromer networks were also characterized for physicochemical properties (swelling, sol fraction, compressive modulus) based on diol length and amount of unsaturated bonds. A statistical model was built using data generated from these diol-based macromers and macromer networks to evaluate the impact of monomer inputs on final macromer and macromer network properties. With the ability to be modified by free radical addition, biodegradable unsaturated polyesters serve as important macromers in the design of devices such as drug delivery vehicles and tissue scaffolds. Given the ability to extensively control final macromer properties based on monomer input parameters, poly(diol fumarate) and poly(diol fumarate-co-succinate) represent an exciting new class of macromers.
Sigma-Aldrich
1,8-Octanediol, 98%