Merck
  • MKRN2 is a novel ubiquitin E3 ligase for the p65 subunit of NF-κB and negatively regulates inflammatory responses.
MKRN2 is a novel ubiquitin E3 ligase for the p65 subunit of NF-κB and negatively regulates inflammatory responses.
Scientific reports (2017-04-06)
Chanyoung Shin, Yuma Ito, Shota Ichikawa, Makio Tokunaga, Kumiko Sakata-Sogawa, Takashi Tanaka
ABSTRACT
Activation of NF-κB transcription factor is strictly regulated to prevent excessive inflammatory responses leading to immunopathology. However, it still remains unclear how NF-κB activation is negatively controlled. The PDZ-LIM domain-containing protein PDLIM2 is a nuclear ubiquitin E3 ligase targeting the p65 subunit of NF-κB for degradation, thus terminating NF-κB-mediated inflammation. Using yeast two-hybrid screening, we sought to isolate PDLIM2-interacting proteins that are critical for suppressing NF-κB signaling. Here we identified MKRN2, a RING finger domain-containing protein that belongs to the makorin ring finger protein gene family, as a novel p65 ubiquitin E3 ligase. MKRN2 bound to p65 and promoted the polyubiquitination and proteasome-dependent degradation of p65 through the MKRN2 RING finger domain, thereby suppressing p65-mediated NF-κB transactivation. Notably, MKRN2 and PDLIM2 synergistically promote polyubiquitination and degradation of p65. Consistently, MKRN2 knockdown in dendritic cells resulted in larger amounts of nuclear p65 and augmented production of proinflammatory cytokines in responses to innate stimuli. These results delineate a novel role of MKRN2 in negatively regulating NF-κB-mediated inflammatory responses, cooperatively with PDLIM2.
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by gel-filtration chromatography