- A selective CB2R agonist (JWH133) restores neuronal circuit after Germinal Matrix Hemorrhage in the preterm via CX3CR1
A selective CB2R agonist (JWH133) restores neuronal circuit after Germinal Matrix Hemorrhage in the preterm via CX3CR1
Neuropharmacology (2017-02-06)
Jun Tang, Hongping Miao, Bing Jiang, Qianwei Chen, Liang Tan, Yihao Tao, Jianbo Zhang, Fabao Gao, Hua Feng, Gang Zhu, Zhi Chen
PMID28153531
ABSTRACT
Microglia play dual roles after germinal matrix hemorrhage, and the neurotrophic phenotype maybe neuroprotective. However, the phenotype transformation and the way by which neuron-microglia dialogue remain unclear. We raise the hypothesis that a cannabinoid receptor2 agonist (JWH133) accelerates the CX3CR1
MATERIALS