Merck
  • Down-regulation of miR-192-5p protects from oxidative stress-induced acute liver injury.
Down-regulation of miR-192-5p protects from oxidative stress-induced acute liver injury.
Clinical science (London, England : 1979) (2016-04-30)
Sanchari Roy, Fabian Benz, Jan Alder, Heike Bantel, Joern Janssen, Mihael Vucur, Jeremie Gautheron, Anne Schneider, Florian Schüller, Sven Loosen, Mark Luedde, Alexander Koch, Frank Tacke, Tom Luedde, Christian Trautwein, Christoph Roderburg
ABSTRACT
miR-192-5p has gained increasing relevance in various diseases, however, its function in acute liver injury is currently unknown. We analysed miR-192-5p serum levels and hepatic miR-192-5p expression in mice after hepatic ischaemia and reperfusion (I/R) as well as in toxic liver injury. On a functional level, miRNA levels were analysed in the different hepatic cell-compartments and in the context of tumour necrosis factor (TNF)-dependent liver cell death. We detected increased serum levels of miR-192-5p after hepatic I/R- and carbon tetrachloride (CCl4)-induced liver injury. miR-192-5p levels correlated with the degree of liver damage and the presence of hepatic cell death detected by TUNEL stainings (terminal deoxynucleotidyltransferase-mediated dUTP biotin nick-end labelling stainings). Moreover, expression of miR-192-5p was increased in a hypoxia/reoxygenation (H/R) model of in vitro hepatocyte injury, supporting that the passive release of miR-192-5p represents a surrogate for hepatocyte death in liver injury. In critically ill patients, miR-192-5p levels were elevated selectively in patients with liver injury and closely correlated with the presence of hepatic injury. In contrast with up-regulated miR-192-5p in the serum, we detected a down-regulation of miR-192-5p in both injured mouse and human livers. Deregulation of miR-192-5p in livers was dependent on stimulation with TNF. Functional experiments confirmed a protective effect of down-regulation of miR-192-5p in hepatocytes, suggesting a role of miR-192-5p in limiting liver injury. Finally, we identified Zeb2, an important regulator of cell death, as a potential target gene mediating the function of miR-192-5p Our data suggest that miR-192-5p is involved in the regulation of liver cell death during acute liver injury and might represent a potent marker of hepatic injury.
Sigma-Aldrich
MISSION® esiRNA, targeting human ZEB2