Merck
  • GFRA1 promotes cisplatin-induced chemoresistance in osteosarcoma by inducing autophagy.
GFRA1 promotes cisplatin-induced chemoresistance in osteosarcoma by inducing autophagy.
Autophagy (2016-10-19)
Mihwa Kim, Ji-Yeon Jung, Seungho Choi, Hyunseung Lee, Liza D Morales, Jeong-Tae Koh, Sun Hun Kim, Yoo-Duk Choi, Chan Choi, Thomas J Slaga, Won Jae Kim, Dae Joon Kim
ABSTRACT
Recent progress in chemotherapy has significantly increased its efficacy, yet the development of chemoresistance remains a major drawback. In this study, we show that GFRA1/GFRα1 (GDNF family receptor α 1), contributes to cisplatin-induced chemoresistance by regulating autophagy in osteosarcoma. We demonstrate that cisplatin treatment induced GFRA1 expression in human osteosarcoma cells. Induction of GFRA1 expression reduced cisplatin-induced apoptotic cell death and it significantly increased osteosarcoma cell survival via autophagy. GFRA1 regulates AMPK-dependent autophagy by promoting SRC phosphorylation independent of proto-oncogene RET kinase. Cisplatin-resistant osteosarcoma cells showed NFKB1/NFκB-mediated GFRA1 expression. GFRA1 expression promoted tumor formation and growth in mouse xenograft models and inhibition of autophagy in a GFRA1-expressing xenograft mouse model during cisplatin treatment effectively reduced tumor growth and increased survival. In cisplatin-treated patients, treatment period and metastatic status were associated with GFRA1-mediated autophagy. These findings suggest that GFRA1-mediated autophagy is a promising novel target for overcoming cisplatin resistance in osteosarcoma.
Sigma-Aldrich
MISSION® esiRNA, targeting human APEX1
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
LR white resin, standard
Sigma-Aldrich
ApopTag Plus Peroxidase In Situ Apoptosis Kit, The ApopTag Plus Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells by labeling & detecting DNA strand breaks by the indirect TUNEL method.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human NFKB1
Sigma-Aldrich
MISSION® esiRNA, targeting human BECN1