Merck
  • SMARCA4-inactivating mutations increase sensitivity to Aurora kinase A inhibitor VX-680 in non-small cell lung cancers.
SMARCA4-inactivating mutations increase sensitivity to Aurora kinase A inhibitor VX-680 in non-small cell lung cancers.
Nature communications (2017-01-20)
Vural Tagal, Shuguang Wei, Wei Zhang, Rolf A Brekken, Bruce A Posner, Michael Peyton, Luc Girard, TaeHyun Hwang, David A Wheeler, John D Minna, Michael A White, Adi F Gazdar, Michael G Roth
ABSTRACT
Mutations in the SMARCA4/BRG1 gene resulting in complete loss of its protein (BRG1) occur frequently in non-small cell lung cancer (NSCLC) cells. Currently, no single therapeutic agent has been identified as synthetically lethal with SMARCA4/BRG1 loss. We identify AURKA activity as essential in NSCLC cells lacking SMARCA4/BRG1. In these cells, RNAi-mediated depletion or chemical inhibition of AURKA induces apoptosis and cell death in vitro and in xenograft mouse models. Disc large homologue-associated protein 5 (HURP/DLGAP5), required for AURKA-dependent, centrosome-independent mitotic spindle assembly is essential for the survival and proliferation of SMARCA4/BRG1 mutant but not of SMARCA4/BRG1 wild-type cells. AURKA inhibitors may provide a therapeutic strategy for biomarker-driven clinical studies to treat the NSCLCs harbouring SMARCA4/BRG1-inactivating mutations.
Sigma-Aldrich
Fluoroshield with DAPI, histology mounting medium
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 300
Sigma-Aldrich
VX-680, ≥98% (HPLC)
Sigma-Aldrich
Anti-BRG1 Antibody, clone 3G4, clone 3G4, from rat