Merck
  • Catalpol induces oligodendrocyte precursor cell-mediated remyelination in vitro.
Catalpol induces oligodendrocyte precursor cell-mediated remyelination in vitro.
American journal of translational research (2016-01-26)
Chun-Xiao Yuan, Takho Chu, Li Liu, Hao-Wen Li, Ya-Jie Wang, An-Chen Guo, Yong-Ping Fan
ABSTRACT
In demyelinating diseases such as multiple sclerosis, one of the treatment strategies includes remyelination using oligodendrocyte precursor cells (OPC). Catalpol, the extract of radix rehmanniae, is neuroprotective. Using an OPC culture model, we showed that 10 μM catalpol promotes OPC proliferation, cell migration and differentiation into mature oligodendrocytes. The 10 μM catalpol displayed stronger effects on OPCs migration and oligodendrocyte differentiation. These results suggest that catalpol has a potential role in promoting remyelination in demyelinating diseases, and is of therapeutic interest.
Product Description
Roche
Anti-Bromodeoxyuridine, from mouse IgG1 (clone: BMC9318)
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse