Merck
  • Experimental challenge of pregnant cattle with the putative abortifacient Waddlia chondrophila.
Experimental challenge of pregnant cattle with the putative abortifacient Waddlia chondrophila.
Scientific reports (2016-11-15)
Nicholas Wheelhouse, Allen Flockhart, Kevin Aitchison, Morag Livingstone, Jeanie Finlayson, Virginie Flachon, Eric Sellal, Mark P Dagleish, David Longbottom
ABSTRACT
Waddlia chondrophila is a Gram-negative intracellular bacterial organism that is related to classical chlamydial species and has been implicated as a cause of abortion in cattle. Despite an increasing number of observational studies linking W. chondrophila infection to cattle abortion, little direct experimental evidence exists. Given this paucity of direct evidence the current study was carried out to investigate whether experimental challenge of pregnant cattle with W. chondrophila would result in infection and abortion. Nine pregnant Friesian-Holstein heifers received 2 × 108 inclusion forming units (IFU) W. chondrophila intravenously on day 105-110 of pregnancy, while four negative-control animals underwent mock challenge. Only one of the challenged animals showed pathogen-associated lesions, with the organism being detected in the diseased placenta. Importantly, the organism was re-isolated and its identity confirmed by whole genome sequencing, confirming Koch's third and fourth postulates. However, while infection of the placenta was observed, the experimental challenge in this study did not confirm the abortifacient potential of the organism.
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule
Sigma-Aldrich
Anti-Bovine IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution