Merck
  • Multiple nanoemulsion system for an oral combinational delivery of oxaliplatin and 5-fluorouracil: preparation and in vivo evaluation.
Multiple nanoemulsion system for an oral combinational delivery of oxaliplatin and 5-fluorouracil: preparation and in vivo evaluation.
International journal of nanomedicine (2016-12-13)
Rudra Pangeni, Sang Won Choi, Ok-Cheol Jeon, Youngro Byun, Jin Woo Park
ABSTRACT
Oxaliplatin (OXA) is a third-generation cisplatin analog that has been approved as first-line chemotherapy in combination with 5-fluorouracil (5-FU) for the treatment of resectable and advanced colorectal cancer. However, the therapeutic efficacy of oral OXA and 5-FU is limited by their low bioavailability due to poor membrane permeability. The aim of the present study was to develop an oral delivery system for OXA and 5-FU. We constructed an ion-pairing complex of OXA with a deoxycholic acid derivative (N
Sigma-Aldrich
(S)-Methyl 6-amino-2-((tert-butoxycarbonyl)amino)hexanoate hydrochloride