Merck
  • Protein kinase Ciota is required for Ras transformation and colon carcinogenesis in vivo.
Protein kinase Ciota is required for Ras transformation and colon carcinogenesis in vivo.
The Journal of cell biology (2004-03-17)
Nicole R Murray, Lee Jamieson, Wangsheng Yu, Jie Zhang, Yesim Gökmen-Polar, Deborah Sier, Panos Anastasiadis, Zoran Gatalica, E Aubrey Thompson, Alan P Fields
ABSTRACT
Protein kinase C iota (PKCiota) has been implicated in Ras signaling, however, a role for PKCiota in oncogenic Ras-mediated transformation has not been established. Here, we show that PKCiota is a critical downstream effector of oncogenic Ras in the colonic epithelium. Transgenic mice expressing constitutively active PKCiota in the colon are highly susceptible to carcinogen-induced colon carcinogenesis, whereas mice expressing kinase-deficient PKCiota (kdPKCiota) are resistant to both carcinogen- and oncogenic Ras-mediated carcinogenesis. Expression of kdPKCiota in Ras-transformed rat intestinal epithelial cells blocks oncogenic Ras-mediated activation of Rac1, cellular invasion, and anchorage-independent growth. Constitutively active Rac1 (RacV12) restores invasiveness and anchorage-independent growth in Ras-transformed rat intestinal epithelial cells expressing kdPKCiota. Our data demonstrate that PKCiota is required for oncogenic Ras- and carcinogen-mediated colon carcinogenesis in vivo and define a procarcinogenic signaling axis consisting of Ras, PKCiota, and Rac1.
Sigma-Aldrich
PKCι, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution