Merck
  • Phosphodiesterase 3B (PDE3B) regulates NLRP3 inflammasome in adipose tissue.
Phosphodiesterase 3B (PDE3B) regulates NLRP3 inflammasome in adipose tissue.
Scientific reports (2016-06-21)
Faiyaz Ahmad, Youn Wook Chung, Yan Tang, Steven C Hockman, Shiwei Liu, Yusuf Khan, Kevin Huo, Eric Billings, Marcelo J Amar, Alan T Remaley, Vincent C Manganiello
ABSTRACT
Activation of inflammation in white adipose tissue (WAT), includes infiltration/expansion of WAT macrophages, contributes pathogenesis of obesity, insulin resistance, and metabolic syndrome. The inflammasome comprises an intracellular sensor (NLR), caspase-1 and the adaptor ASC. Inflammasome activation leads to maturation of caspase-1 and processing of IL1β, contributing to many metabolic disorders and directing adipocytes to a more insulin-resistant phenotype. Ablation of PDE3B in WAT prevents inflammasome activation by reducing expression of NLRP3, caspase-1, ASC, AIM2, TNFα, IL1β and proinflammatory genes. Following IP injection of lipopolysaccharide (LPS), serum levels of IL1β and TNFα were reduced in PDE3B(-/-)mice compared to WT. Activation of signaling cascades, which mediate inflammasome responses, were modulated in PDE3B(-/-)mice WAT, including smad, NFAT, NFkB, and MAP kinases. Moreover, expression of chemokine CCL2, MCP-1 and its receptor CCR2, which play an important role in macrophage chemotaxis, were reduced in WAT of PDE3B(-/-)mice. In addition, atherosclerotic plaque formation was significantly reduced in the aorta of apoE(-/-)/PDE3B(-/-)and LDL-R(-/-)/PDE3B(-/-)mice compared to apoE(-/-)and LDL-R(-/-)mice, respectively. Obesity-induced changes in serum-cholesterol were blocked in PDE3B(-/-)mice. Collectively, these data establish a role for PDE3B in modulating inflammatory response, which may contribute to a reduced inflammatory state in adipose tissue.
Sigma-Aldrich
Anti-IKKα Mouse mAb (14A231), liquid, clone 14A231, Calbiochem®
Sigma-Aldrich
Cilostamide, A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase (PDE III; IC₅₀ = 70 nM).