Merck
  • Novel method of aromatic coupling between N-aryl methanesulfonamide and thiophene derivatives.
Novel method of aromatic coupling between N-aryl methanesulfonamide and thiophene derivatives.
Organic letters (2007-05-29)
Alexandre Jean, Jonathan Cantat, Didier Bérard, Denis Bouchu, Sylvain Canesi
ABSTRACT
Oxidation of N-aromatic methanesulfonamides with iodobenzene diacetate in the presence of substituted thiophene promotes interesting coupling reactions in moderate to good yields.
Sigma-Aldrich
3-Bromothiophene, 97%