Merck
  • miR-17-92/p38α Dysregulation Enhances Wnt Signaling and Selects Lgr6+ Cancer Stem-like Cells during Lung Adenocarcinoma Progression.
miR-17-92/p38α Dysregulation Enhances Wnt Signaling and Selects Lgr6+ Cancer Stem-like Cells during Lung Adenocarcinoma Progression.
Cancer research (2016-05-20)
Anna Guinot, Feride Oeztuerk-Winder, Juan-Jose Ventura
ABSTRACT
Defining the molecular and cellular roots of lung cancer relapse after initial treatment remains an imperative to improve survival. Here we report that the lung stem cell marker Lgr6 becomes enriched in non-small cell lung cancer (NSCLC) cells during malignant progression. Lgr6(+) NSCLC cells displayed self-renewal and differentiation properties along with a higher tumorigenic potential. Mechanistic investigations suggested that a defective repression of the miR-17-92 gene cluster was responsible for evolution of a selection for outgrowth of Lgr6(+) NSCLC cells. High levels of expression of miR-19 family members were found to target and downregulate levels of p38α kinase, providing a specific survival signal for Lgr6(+) cells as mediated by increased Wnt/ß-catenin activity. Our results identify a specific stem-like cell population in NSCLC with increased malignant potential, the elucidation of which may enable earlier prognosis and possibly the development of more effective targeted treatments. Cancer Res; 76(13); 4012-22. ©2016 AACR.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Roche
Red Blood Cell Lysis Buffer, solution, Roche, pkg of 100 mL, sufficient for 50-500 reactions