Merck
  • CRL4(Wdr70) regulates H2B monoubiquitination and facilitates Exo1-dependent resection.
CRL4(Wdr70) regulates H2B monoubiquitination and facilitates Exo1-dependent resection.
Nature communications (2016-04-22)
Ming Zeng, Laifeng Ren, Ken'Ichi Mizuno, Konstantinos Nestoras, Haibin Wang, Zizhi Tang, Liandi Guo, Daochun Kong, Qiwen Hu, Qun He, Lilin Du, Antony M Carr, Cong Liu
ABSTRACT
Double-strand breaks repaired by homologous recombination (HR) are first resected to form single-stranded DNA, which binds replication protein A (RPA). RPA attracts mediators that load the Rad51 filament to promote strand invasion, the defining feature of HR. How the resection machinery navigates nucleosome-packaged DNA is poorly understood. Here we report that in Schizosaccharomyces pombe a conserved DDB1-CUL4-associated factor (DCAF), Wdr70, is recruited to DSBs as part of the Cullin4-DDB1 ubiquitin ligase (CRL4(Wdr70)) and stimulates distal H2B lysine 119 mono-ubiquitination (uH2B). Wdr70 deletion, or uH2B loss, results in increased loading of the checkpoint adaptor and resection inhibitor Crb2(53BP1), decreased Exo1 association and delayed resection. Wdr70 is dispensable for resection upon Crb2(53BP1) loss, or when the Set9 methyltransferase that creates docking sites for Crb2 is deleted. Finally, we establish that this histone regulatory cascade similarly controls DSB resection in human cells.
Product Description
Roche
High Pure PCR Template Preparation Kit, pkg of 100 purifications, suitable for DNA extraction
Roche
X-tremeGENE HP DNA Transfection Reagent, High-performance polymer reagent for transfecting many cell lines
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial