Merck
  • Synthesis and Anti-inflammatory Performance of Newly Cyclizine Derivatives on Adult Male Wistar Rats.
Synthesis and Anti-inflammatory Performance of Newly Cyclizine Derivatives on Adult Male Wistar Rats.
Iranian journal of pharmaceutical research : IJPR (2013-11-20)
Abbas Ahmadi, Mohsen Khalili, Shahnaz Chavrogh, Babak Nahri-Niknafs
ABSTRACT
Cyclizine (1-benzhydryl-4-methyl-piperazine, CAS 82-92-8, CYC, I), a piperazine derivative, belongs to H1 antihistamine group of drugs that shows such pharmacological properties as anti-inflammatory, anti-allergic and anti-platelet effects, similar to other H1-receptor antagonists. In this study, two new tolyl and cumene derivatives of I (1-ethyl- 4-[(p-isopropylphenyl) (p-tolyl) methyl]-piperazine, II and 1-[3, 4-dichlorophenyl]-4-[[p-isopropylphenyl] [p-tolyl] methyl]-piperazine, III) were synthesized to investigate their acute and chronic anti-inflammatory activities in formalin and histamine-induced rat paw edema. In addition, the vascular permeability in formalin and histamine-induced paw edema, xylene-induced ear edema, and peritonitis due to acetic acid application into peritoneal cavity were measured. The cotton pellet-induced granuloma model was chosen for inducing chronic inflammation in rats. Findings proved reduction in formalin-induced rat paw edema and vascular permeability (acute inflammation) by I and II at 30 min after the injection. In addition, results in histamine-induced rat paw edema showed anti-inflammatory effects of all drugs started 60 min after the injection as these effects continued for a longer period by II and III comparing to I, as discussed above. In addition, the data on vascular permeability in xylene-induced ear edema and acetic acid-induced to peritoneal cavity confirmed that substitutions on cyclizine molecule were more effective and could decrease the vascular permeability and acute inflammation. However, the results from the cotton pellet-induced granuloma formation in rats revealed that none of the drugs (I-III) were effective to reduce the reactions and intermediates of chronic inflammation.