Merck
  • Quorum sensing and Chromobacterium violaceum: exploitation of violacein production and inhibition for the detection of N-acylhomoserine lactones.
Quorum sensing and Chromobacterium violaceum: exploitation of violacein production and inhibition for the detection of N-acylhomoserine lactones.
Microbiology (Reading, England) (1998-01-09)
K H McClean, M K Winson, L Fish, A Taylor, S R Chhabra, M Camara, M Daykin, J H Lamb, S Swift, B W Bycroft, G S Stewart, P Williams
ABSTRACT
Quorum sensing relies upon the interaction of a diffusible signal molecule with a transcriptional activator protein to couple gene expression with cell population density. In Gram-negative bacteria, such signal molecules are usually N-acylhomoserine lactones (AHLs) which differ in the structure of their N-acyl side chains. Chromobacterium violaceum, a Gram-negative bacterium commonly found in soil and water, produces the characteristic purple pigment violacein. Previously the authors described a violacein-negative, mini-Tn5 mutant of C. violaceum (CV026) in which pigment production can be restored by incubation with supernatants from the wild-type strain. To develop this mutant as a general biosensor for AHLs, the natural C. violaceum AHL molecule was first chemically characterized. By using solvent extraction, HPLC and mass spectrometry, a single AHL, N-hexanoyl-L-homoserine lactone (HHL), was identified in wild-type C. violaceum culture supernatants which was absent from CV026. Since the production of violacein constitutes a simple assay for the detection of AHLs, we explored the ability of CV026 to respond to a series of synthetic AHL and N-acylhomocysteine thiolactone (AHT) analogues. In CV026, violacein is inducible by all the AHL and AHT compounds evaluated with N-acyl side chains from C4 to C8 in length, with varying degrees of sensitivity. Although AHL compounds with N-acyl side chains from C10 to C14 are unable to induce violacein production, if an activating AHL (e.g. HHL) is incorporated into the agar, these long-chain AHLs can be detected by their ability to inhibit violacein production. The versatility of CV026 in facilitating detection of AHL mixtures extracted from culture supernatants and separated by thin-layer chromatography is also demonstrated. These simple bioassays employing CV026 thus greatly extend the ability to detect a wide spectrum of AHL signal molecules.
Sigma-Aldrich
N-Hexanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Decanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Octanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Heptanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Tetradecanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Octanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Butyryl-DL-homoserine lactone, ≥96.0% (HPLC)
Sigma-Aldrich
N-Dodecanoyl-DL-homoserine lactone, ≥97.0% (HPLC)