Merck
  • Broadening the versatility of lentiviral vectors as a tool in nucleic acid research via genetic code expansion.
Broadening the versatility of lentiviral vectors as a tool in nucleic acid research via genetic code expansion.
Nucleic acids research (2015-03-15)
Yongxiang Zheng, Fei Yu, Yiming Wu, Longlong Si, Huan Xu, Chuanling Zhang, Qing Xia, Sulong Xiao, Qi Wang, Qiuchen He, Peng Chen, Jiangyun Wang, Kazunari Taira, Lihe Zhang, Demin Zhou
ABSTRACT
With the aim of broadening the versatility of lentiviral vectors as a tool in nucleic acid research, we expanded the genetic code in the propagation of lentiviral vectors for site-specific incorporation of chemical moieties with unique properties. Through systematic exploration of the structure-function relationship of lentiviral VSVg envelope by site-specific mutagenesis and incorporation of residues displaying azide- and diazirine-moieties, the modifiable sites on the vector surface were identified, with most at the PH domain that neither affects the expression of envelope protein nor propagation or infectivity of the progeny virus. Furthermore, via the incorporation of such chemical moieties, a variety of fluorescence probes, ligands, PEG and other functional molecules are conjugated, orthogonally and stoichiometrically, to the lentiviral vector. Using this methodology, a facile platform is established that is useful for tracking virus movement, targeting gene delivery and detecting virus-host interactions. This study may provide a new direction for rational design of lentiviral vectors, with significant impact on both basic research and therapeutic applications.
Sigma-Aldrich
N6-((2-Azidoethoxy)carbonyl)-L-lysine hydrochloride, ≥95%
Sigma-Aldrich
Monoclonal Anti-VSV Glycoprotein antibody produced in mouse, clone P5D4, ascites fluid