Merck
  • A qualitative evaluation of 40 voluntary, smoke-free, multiunit, housing policy campaigns in California.
A qualitative evaluation of 40 voluntary, smoke-free, multiunit, housing policy campaigns in California.
Tobacco control (2013-06-21)
Travis D Satterlund, Jeanette Treiber, Robin Kipke, Diana Cassady
ABSTRACT
Although it is legal for multiunit housing (MUH) property owners in all 50 states to prohibit smoking on their premises, including in individual units, MUH constitutes a relatively new setting to reduce exposure to secondhand smoke via voluntary smoke-free policy. This paper examines California state-funded smoke-free MUH policy campaigns between 2004 and 2010. A cross-case analysis of 40 state-funded smoke-free MUH policy campaigns was conducted via an examination of final evaluation reports submitted to the California Tobacco Control Program. The most effective voluntary smoke-free MUH policy campaigns typically included: (1) learning the local [MUH] context, (2) finding and using a champion, (3) partnering with like-minded organisations, (4) building relationships with stakeholders, (5) collecting and using local data and (6) making a compelling case to decision makers. The aforementioned steps tended to be intertwined, and successfully securing voluntary smoke-free MUH policy required a strategic but flexible plan of implementation prior to entrance into the field. Campaigns designed to enhance voluntary smoke-free MUH policy adoption should underscore the economic viability of such policies during each strategic step.