Merck
  • Expedient protocol for solid-phase synthesis of secondary and tertiary amines.
Expedient protocol for solid-phase synthesis of secondary and tertiary amines.
Organic letters (2004-06-05)
Christian A Olsen, Matthias Witt, Jerzy W Jaroszewski, Henrik Franzyk
ABSTRACT
[reaction: see text] An expedient solid-phase synthetic approach to secondary and tertiary amines was developed. The protocol employs conversion of resin-bound amino alcohols to the corresponding iodides, followed by iodide displacement with primary or secondary amines or with unprotected amino alcohols. This two-step procedure, affording products in good to excellent yields, is suitable for solid-phase synthesis of polyamines.
Sigma-Aldrich
4-Piperidinemethanol, 97%