Merck
  • Identification of a novel de novo mutation of CREBBP in a patient with Rubinstein-Taybi syndrome by targeted next-generation sequencing: a case report.
Identification of a novel de novo mutation of CREBBP in a patient with Rubinstein-Taybi syndrome by targeted next-generation sequencing: a case report.
Human pathology (2015-11-26)
Kunka Kamenarova, Emil Simeonov, Reni Tzveova, Daniela Dacheva, Marin Penkov, Ivo Kremensky, Penka Perenovska, Vanio Mitev, Radka Kaneva
ABSTRACT
Rubinstein-Taybi syndrome (RSTS) is a rare autosomal dominant congenital disorder (prevalence, 1:125000-720000) characterized by broad thumbs and halluces, facial dysmorphism, psychomotor development delay, skeletal defects, abnormalities in the posterior fossa, and short stature. The purpose of this study was to use targeted exome sequencing to identify the genetic cause of RSTS in a 6.5-year-old girl presenting typical features of this condition. Targeted exome sequencing of the patient DNA revealed de novo transition c.1066C>T corresponding to a novel nonsense mutation p.Q356X in the CREB-binding protein gene, CREBBP, whose haploinsufficiency is responsible for 50% to 60% of the RSTS cases. Based on comparing the clinical manifestations of our patient with those of patients carrying similar mutations, we supposed that haploinsufficiency is the possible functional consequence of p.Q356X mutation by creation of a loss-of-function CREBBP allele due to a premature stop codon and RSTS phenotype. Our findings expand the spectrum of mutations associated with this condition.
Sigma-Aldrich
PF-CBP1, ≥98% (HPLC)