Merck
  • Improved Treatment of Breast Cancer with Anti-HER2 Therapy Requires Interleukin-21 Signaling in CD8+ T Cells.
Improved Treatment of Breast Cancer with Anti-HER2 Therapy Requires Interleukin-21 Signaling in CD8+ T Cells.
Cancer research (2016-01-09)
Deepak Mittal, Franco Caramia, Stefan Michiels, Heikki Joensuu, Pirkko-Liisa Kellokumpu-Lehtinen, Christos Sotiriou, Sherene Loi, Mark J Smyth
ABSTRACT
The HER2/ErbB2 monoclonal antibody (mAb) trastuzumab is combined with chemotherapy as a standard-of-care for newly diagnosed HER2(+) breast cancer patients, but some patients treated with this combination therapy experience early relapse. Our analysis of data from a clinical trial evaluating the efficacy of chemotherapy plus/minus trastuzumab suggested that the magnitude of trastuzumab benefit on distant disease-free survival was higher for increasing expression of the IL21 receptor (IL21R). Therefore, we investigated a possible role for IL21 signaling in promoting HER2 mAb therapeutic efficacy. We found that IL21R-deficient mice and wild-type mice treated with a neutralizing anti-IL21 mAb were less susceptible to trastuzumab-like anti-ErbB2 therapy. Furthermore, IL21R expression on CD8(+) T cells, but not on natural killer cells, was required for optimal anti-ErbB2 mAb efficacy, and IL21 expression was enhanced in tumor-infiltrating CD4(+) T lymphocytes after anti-ErbB2 therapy. Finally, we found that administering recombinant IL21 in combination with anti-ErbB2 therapy was therapeutic against primary tumors and experimental metastases in mice. Collectively, our findings suggest that elevating IL21 signaling may enhance trastuzumab efficacy, thus constituting a novel candidate strategy to overcome trastuzumab resistance and improve patient survival. Cancer
Sigma-Aldrich
Interleukin-21 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture