Merck
Nuclear TRAF3 is a negative regulator of CREB in B cells.
Proceedings of the National Academy of Sciences of the United States of America (2016-01-13)
Nurbek Mambetsariev, Wai W Lin, Laura L Stunz, Brett M Hanson, Joanne M Hildebrand, Gail A Bishop
ABSTRACT
The adaptor protein TNF receptor-associated factor 3 (TRAF3) regulates signaling through B-lymphocyte receptors, including CD40, BAFF receptor, and Toll-like receptors, and also plays a critical role inhibiting B-cell homoeostatic survival. Consistent with these findings, loss-of-function human TRAF3 mutations are common in B-cell cancers, particularly multiple myeloma and B-cell lymphoma. B cells of B-cell-specific TRAF3(-/-) mice (B-Traf3(-/-)) display remarkably enhanced survival compared with littermate control (WT) B cells. The mechanism for this abnormal homeostatic survival is poorly understood, a key knowledge gap in selecting optimal treatments for human B-cell cancers with TRAF3 deficiency. We show here for the first time to our knowledge that TRAF3 is a resident nuclear protein that associates with the transcriptional regulator cAMP response element binding protein (CREB) in both mouse and human B cells. The TRAF-C domain of TRAF3 was necessary and sufficient to localize TRAF3 to the nucleus via a functional nuclear localization signal. CREB protein was elevated in TRAF3(-/-) B cells, without change in mRNA, but with a decrease in CREB ubiquitination. CREB-mediated transcriptional activity was increased in TRAF3-deficient B cells. Consistent with these findings, Mcl-1, an antiapoptotic target of CREB-mediated transcription, was increased in the absence of TRAF3 and enhanced Mcl-1 was suppressed with CREB inhibition. TRAF3-deficient B cells were also preferentially sensitive to survival inhibition with pharmacologic CREB inhibitor. Our results identify a new mechanism by which nuclear TRAF3 regulates B-cell survival via inhibition of CREB stability, information highly relevant to the role of TRAF3 in B-cell malignancies.
Sigma-Aldrich
Naphthol AS-E phosphate, suitable for histology