Merck
  • Single cell imaging of Bruton's tyrosine kinase using an irreversible inhibitor.
Single cell imaging of Bruton's tyrosine kinase using an irreversible inhibitor.
Scientific reports (2014-04-25)
Anna Turetsky, Eunha Kim, Rainer H Kohler, Miles A Miller, Ralph Weissleder
ABSTRACT
A number of Bruton's tyrosine kinase (BTK) inhibitors are currently in development, yet it has been difficult to visualize BTK expression and pharmacological inhibition in vivo in real time. We synthesized a fluorescent, irreversible BTK binder based on the drug Ibrutinib and characterized its behavior in cells and in vivo. We show a 200 nM affinity of the imaging agent, high selectivity, and irreversible binding to its target following initial washout, resulting in surprisingly high target-to-background ratios. In vivo, the imaging agent rapidly distributed to BTK expressing tumor cells, but also to BTK-positive tumor-associated host cells.
Sigma-Aldrich
N-Boc-2-aminoacetaldehyde, 95%