Merck
  • Preparation, quality control and biodistribution assessment of ¹⁵³Sm-BPAMD as a novel agent for bone pain palliation therapy.
Preparation, quality control and biodistribution assessment of ¹⁵³Sm-BPAMD as a novel agent for bone pain palliation therapy.
Annals of nuclear medicine (2015-08-12)
Ali Rabie, Razieh Enayati, Hassan Yousefnia, Amir Reza Jalilian, Mojtaba Shamsaei, Samaneh Zolghadri, Ali Bahrami-Samani, Mohammad Hosntalab
ABSTRACT
Various phosphonate ligands labeled with β(-)-emitting radionuclides have shown good efficacy for bone pain palliation. In this study, a new agent for bone pain palliation has been developed. ¹⁵³Sm-(4-{[(bis(phosphonomethyl))carbamoyl]methyl}-7,10-bis(carboxymethyl)-1,4,7,10-tetraazacyclododec-1-yl) acetic acid (¹⁵³Sm-BPAMD) complex was prepared using BPAMD ligand and ¹⁵³SmCl3. The effect of various parameters on the labeling yield of ¹⁵³Sm-BPAMD including ligand concentration, pH, temperature and reaction time were studied. Radiochemical purity of the radiolabeled complex was checked by instant thin layer chromatography (ITLC). Stability studies of the complex in the final preparation and in the presence of human serum were performed up to 48 h. Partition coefficient and hydroxyapatite (HA) binding of the complex were investigated and biodistribution studies (SPECT imaging and scarification) were performed after injection of the complex to Syrian mice up to 48 h post-injection. The biodistribution of the complex was compared with the biodistribution of the ¹⁵³Sm cation in the same type mice. ¹⁵³Sm-BPAMD was prepared in high radiochemical purity >98% and specific activity of 267 GBq/mmol at the optimal conditions. The complex demonstrated significant stability at room temperature and in human serum at least for 48 h. HA binding assay demonstrated that at the amount of more than 5 mg, approximately, all radiolabeled complex was bound to HA. At the pH 7.4, LogP o/w was -1.86 ± 0.02. Both SPECT and scarification showed major accumulation of the labeled compound in the bone tissue. The results show that ¹⁵³Sm-BPAMD has interesting characteristics as an agent for bone pain palliation; however, further biological studies in other mammals are still needed.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%