Merck
  • An Extended-gate Type Organic FET Based Biosensor for Detecting Biogenic Amines in Aqueous Solution.
An Extended-gate Type Organic FET Based Biosensor for Detecting Biogenic Amines in Aqueous Solution.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-07-15)
Tsuyoshi Minami, Tsubasa Sato, Tsukuru Minamiki, Shizuo Tokito
ABSTRACT
This study is the first to report on the detection of biogenic amines in an aqueous solution using an organic field-effect transistor (OFET) device with an extended gate electrode modified with a layer of diamine oxidase and a horseradish peroxidase osmium-redox polymer. The limit of detection (LOD) for histamine was estimated to be 1.2 μM. These results reveal that extended-gate type OFET devices are highly suitable enzyme-based biosensors for the detection of biogenic amine levels.
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Spermine tetrahydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Spermine tetrahydrochloride, powder or crystals
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Spermine tetrahydrochloride, BioReagent, for molecular biology
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Spermine tetrahydrochloride, ≥99.0% (AT), powder or crystals
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Tetradecylphosphonic acid, 98%
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Sigma-Aldrich
1,2-Dichlorobenzene solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Spermidine, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder