Merck
  • Changes in soymilk during fermentation with kefir culture: oligosaccharides hydrolysis and isoflavone aglycone production.
Changes in soymilk during fermentation with kefir culture: oligosaccharides hydrolysis and isoflavone aglycone production.
International journal of food sciences and nutrition (2015-10-16)
T R Baú, S Garcia, E I Ida
ABSTRACT
The objective of this study was to evaluate the changes in oligosaccharides and isoflavone aglycone content in soymilk during fermentation with commercial kefir culture. Soymilk was fermented with kefir culture at 25 °C for 30 h. The counts of lactic acid bacteria, Lactococcus lactis, Leuconostoc sp and yeasts; measurements of pH, acidity, α-galactosidase and β-glucosidase activity, sugar and isoflavone contents were performed at the intervals of time. In the fermented soymilk, the lactic acid bacteria counts increased from 7.6 log to 9.1 CFU g(-1), pH reached to 4.9 and lactic acid reached 0.34 g 100  g(- 1). The α-galactosidase was produced (0.016 AU g(-1)) with 100% raffinose and 92% stachyose hydrolysis being observed after the depletion of galactose, glucose and sucrose. Kefir culture produced β-glucosidase (0.0164 AU g(-1)), resulting in 100% bioconversion of glycitin and daidzin and 89% bioconversion of genistin into the corresponding aglycones. The fermented soymilk presented 1.67 μmol g(-1) of daidzein, 0.28 μmol g(-1) of glicitein and 1.67 μmol g (-1) of genistein.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)