Merck
  • Inherent organic compounds in biochar--Their content, composition and potential toxic effects.
Inherent organic compounds in biochar--Their content, composition and potential toxic effects.
Journal of environmental management (2015-04-08)
Wolfram Buss, Ondřej Mašek, Margaret Graham, Dominik Wüst
ABSTRACT
Pyrolysis liquids consist of thermal degradation products of biomass in various stages of its decomposition. Therefore, if biochar gets affected by re-condensed pyrolysis liquids it is likely to contain a huge variety of organic compounds. In this study the chemical composition of such compounds associated with two contaminated, high-volatile organic compound (VOC) biochars were investigated and compared with those for a low-VOC biochar. The water-soluble organic compounds with the highest concentrations in the two high-VOC biochars were acetic, formic, butyric and propionic acids; methanol, phenol, o-, m- and p-cresol, and 2,4-dimethylphenol, all with concentrations over 100 μg g(-1). The concentrations of 16 US EPA PAHs determined by 36 h toluene extractions were 6.09 μg g(-1) for the low-VOC biochar. For high-VOC biochar the total concentrations were 53.42 μg g(-1) and 27.89 μg g(-1), while concentrations of water-soluble PAHs ranged from 1.5 to 2 μg g(-1). Despite the concentrations of PAHs exceeding biochar guideline values, it was concluded that, for these particular biochars, the biggest concern for application to soil would be the co-occurrence of VOCs such as low molecular weight (LMW) organic acids and phenols, as these can be highly mobile and have a high potential to cause phytotoxic effects. Therefore, based on results of this study we strongly suggest for VOCs to be included among criteria for assessment of biochar quality.
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Toluene-d8, 99 atom % D
Sigma-Aldrich
Toluene-d8, 99 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Toluene-d8, "100%", 99.96 atom % D
Sigma-Aldrich
Toluene-d8, 99.6 atom % D
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Carbon disulfide, anhydrous, ≥99%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)