Merck
  • Tanshinone IIA - loaded pellets developed for angina chronotherapy: Deconvolution-based formulation design and optimization, pharmacokinetic and pharmacodynamic evaluation.
Tanshinone IIA - loaded pellets developed for angina chronotherapy: Deconvolution-based formulation design and optimization, pharmacokinetic and pharmacodynamic evaluation.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-05-16)
Hong-Xiang Yan, Jin Li, Zheng-Hua Li, Wen-Li Zhang, Jian-Ping Liu
ABSTRACT
This paper put forward a deconvolution-based method for designing and optimizing tanshinone IIA sustained-release pellets (TA-SRPs) with improved efficacy in the treatment of variant angina. TA-SRPs were prepared by coating TA ternary solid dispersion immediate-release pellets (TA-tSD-IRPs) with the blends of polyvinyl acetate (PVAc) and polyvinyl alcohol-polyethylene glycol (PVA-PEG) using fluidized bed technology. The plasma concentration-time curve of TA-tSD-IRPs following oral administration as a weight function was investigated in New Zealand white rabbits. The predicted/expected plasma concentration-time curve of TA-SRPs as a response function was simulated according to the circadian rhythm of variant angina during 24h based on chronotherapy theory. The desired drug release profile of TA-SRPs was obtained via the point-area deconvolution procedure using the weight function and response function, and used for formulation optimization of TA-SRPs. The coating formulation of TA-SRPs was optimized as 70:30 (w/w) PVAc/PVA-PEG with 5% (w/w) coating weight due to in vitro drug release profile of these TA-SRPs was similar to the desired release profile (similarity factor f2=64.90). Pharmacokinetic studies of these optimized TA-SRPs validated that their actual plasma concentration-time curve possessed a basically consistent trend with the predicted plasma concentration-time curve and the absolute percent errors (%PE) of concentrations in 8-12h were less than 10%. Pharmacodynamic studies further demonstrated that these TA-SRPs had stable and improved efficacy with almost simultaneous drug concentration-efficacy. In conclusion, deconvolution could be employed in the development of TA-SRPs for angina chronotherapy with simultaneous drug efficacy and reduced design blindness and complexity.
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture