Merck
  • Magnetic, fluorescent, and thermo-responsive poly(MMA-NIPAM-Tb(AA)3Phen)/Fe3O4 multifunctional nanospheres prepared by emulsifier-free emulsion polymerization.
Magnetic, fluorescent, and thermo-responsive poly(MMA-NIPAM-Tb(AA)3Phen)/Fe3O4 multifunctional nanospheres prepared by emulsifier-free emulsion polymerization.
Journal of biomaterials applications (2015-03-15)
Ying Gong, Jingwen Dai, Huan Li, Xin Wang, Haoran Xiong, Quanyuan Zhang, Penghui Li, Changfeng Yi, Zushun Xu, Haibo Xu, Paul K Chu
ABSTRACT
Magnetic, luminescent, and thermoresponsive multifunctional nanospheres composed of modified Fe3O4 nanoparticles as the core and rare earth complex Tb(AA)3Phen as the shell are synthesized by emulsifier-free emulsion polymerization. The core-shell spherical structure has a size between 140 and 220 nm and exhibits strong green fluorescence of the rare earth complex Tb(AA)3Phen. In the R2 relaxivity and in vivo MRI studies, the R2 relaxivity of the nanospheres is 562.56 mM(-1) s(-1) and enhanced T2-weighted images are observed from the nanospheres in the liver and spleen after injection as a contrast agent. The excellent superparamagnetic, thermosensitive, and fluorescent properties render the nanospheres useful in biomedical engineering and optical imaging.
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)