Merck
  • mTOR Complex Signaling through the SEMA4A-Plexin B2 Axis Is Required for Optimal Activation and Differentiation of CD8+ T Cells.
mTOR Complex Signaling through the SEMA4A-Plexin B2 Axis Is Required for Optimal Activation and Differentiation of CD8+ T Cells.
Journal of immunology (Baltimore, Md. : 1950) (2015-06-28)
Daisuke Ito, Satoshi Nojima, Masayuki Nishide, Tatsusada Okuno, Hyota Takamatsu, Sujin Kang, Tetsuya Kimura, Yuji Yoshida, Keiko Morimoto, Yohei Maeda, Takashi Hosokawa, Toshihiko Toyofuku, Jun Ohshima, Daisuke Kamimura, Masahiro Yamamoto, Masaaki Murakami, Eiichi Morii, Hiromi Rakugi, Yoshitaka Isaka, Atsushi Kumanogoh
ABSTRACT
Mammalian target of rapamycin (mTOR) plays crucial roles in activation and differentiation of diverse types of immune cells. Although several lines of evidence have demonstrated the importance of mTOR-mediated signals in CD4(+) T cell responses, the involvement of mTOR in CD8(+) T cell responses is not fully understood. In this study, we show that a class IV semaphorin, SEMA4A, regulates CD8(+) T cell activation and differentiation through activation of mTOR complex (mTORC) 1. SEMA4A(-/-) CD8(+) T cells exhibited impairments in production of IFN-γ and TNF-α and induction of the effector molecules granzyme B, perforin, and FAS-L. Upon infection with OVA-expressing Listeria monocytogenes, pathogen-specific effector CD8(+) T cell responses were significantly impaired in SEMA4A(-/-) mice. Furthermore, SEMA4A(-/-) CD8(+) T cells exhibited reduced mTORC1 activity and elevated mTORC2 activity, suggesting that SEMA4A is required for optimal activation of mTORC1 in CD8(+) T cells. IFN-γ production and mTORC1 activity in SEMA4A(-/-) CD8(+) T cells were restored by administration of recombinant Sema4A protein. In addition, we show that plexin B2 is a functional receptor of SEMA4A in CD8(+) T cells. Collectively, these results not only demonstrate the role of SEMA4A in CD8(+) T cells, but also reveal a novel link between a semaphorin and mTOR signaling.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Plxnb2
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PLXNB2
SAFC
BIS-TRIS
SAFC
BIS-TRIS