Merck
  • Accumulation of polycyclic aromatic hydrocarbons by lichen transplants: Comparison with gas-phase passive air samplers.
Accumulation of polycyclic aromatic hydrocarbons by lichen transplants: Comparison with gas-phase passive air samplers.
Chemosphere (2015-04-26)
S Loppi, K Pozo, V H Estellano, S Corsolini, G Sardella, L Paoli
ABSTRACT
This study compared the accumulation of 16 polycyclic aromatic hydrocarbons (PAHs) in samples of the lichen Evernia prunastri exposed for 3 months in and around an industrial area of S Italy with that in co-located passive gas-phase air samplers. The results showed a strong linear correlations (R=0.96, P<0.05) between total PAHs in lichens and in passive samplers, clearly indicating that lichen transplants may provide direct quantitative information on the atmospheric load by total PAHs, allowing translation of lichen values into atmospheric concentrations. To the best of our knowledge this is the first study reporting such a correlation with gas-phase passive air samplers.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Benzo[k]fluoranthene, for fluorescence, ≥99%
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Acenaphthylene, 99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Benzo[ghi]perylene, 98%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Benzo[b]fluoranthene, 98%
Sigma-Aldrich
Acenaphthylene, 75%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Fluorene, 98%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Sigma-Aldrich
Fluoranthene, 98%
Sigma-Aldrich
Acenaphthene, 99%
Sigma-Aldrich
Phenanthrene-d10, 98 atom % D, 98% (CP)
Sigma-Aldrich
Phenanthrene, 98%
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)