Merck
  • NSun2 Promotes Cell Growth via Elevating Cyclin-Dependent Kinase 1 Translation.
NSun2 Promotes Cell Growth via Elevating Cyclin-Dependent Kinase 1 Translation.
Molecular and cellular biology (2015-09-24)
Junyue Xing, Jie Yi, Xiaoyu Cai, Hao Tang, Zhenyun Liu, Xiaotian Zhang, Jennifer L Martindale, Xiaoling Yang, Bin Jiang, Myriam Gorospe, Wengong Wang
ABSTRACT
The tRNA methytransferase NSun2 promotes cell proliferation, but the molecular mechanism has not been elucidated. Here, we report that NSun2 regulates cyclin-dependent kinase 1 (CDK1) expression in a cell cycle-dependent manner. Knockdown of NSun2 decreased the CDK1 protein level, while overexpression of NSun2 elevated it without altering CDK1 mRNA levels. Further studies revealed that NSun2 methylated CDK1 mRNA in vitro and in cells and that methylation by NSun2 enhanced CDK1 translation. Importantly, NSun2-mediated regulation of CDK1 expression had an impact on the cell division cycle. These results provide new insight into the regulation of CDK1 during the cell division cycle.
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
MISSION® esiRNA, targeting human SPINK5
Sigma-Aldrich
MISSION® esiRNA, targeting human GNL3
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Spink5
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gnl3
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, ≥99.5% (HPLC)