Merck
  • Growth of healthy and sanitizer-injured Salmonella cells on mung bean sprouts in different commercial enrichment broths.
Growth of healthy and sanitizer-injured Salmonella cells on mung bean sprouts in different commercial enrichment broths.
Food microbiology (2015-09-05)
Qianwang Zheng, Marta Mikš-Krajnik, Craig D'Souza, Yishan Yang, Da-Jeong Heo, Si-Kyung Kim, Seung-Cheol Lee, Hyun-Gyun Yuk
ABSTRACT
The ability of nine commercial broths to enrich healthy and 90% sanitizer-injured Salmonella Typhimurium and Salmonella cocktail on mung bean sprouts was evaluated to select an optimum broth for detection. Results showed that S. Typhimurium multiplied faster and reached a higher population in buffered peptone water (BPW), Salmonella AD media (AD) and ONE broth-Salmonella (OB), compared with other broths. Healthy and 90% sanitizer-injured Salmonella at low concentrations increased by 4.0 log CFU/ml in these three broths. However, no Salmonella growth was observed in lactose broth (LB). Further investigation showed that during incubation, pH of LB dropped from 6.7 to 4.2, due to production of lactic (66 mM) and acetic acids (62 mM) by lactic acid bacteria that were identified as dominant microbiota in bean sprouts. Though no cell membrane damage was detected by propidium monoazide combined with real-time PCR, it was found that LB inhibited Salmonella growth, especially from low inoculum levels. This study suggests that in consideration of effectiveness and cost, BPW would be a suitable enrichment broth to use for isolating and detecting Salmonella on mung bean sprouts, while using LB might cause false negative results in Salmonella detection by either PCR or standard cultural method.
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Millipore
Universal Pre-Enrichment Broth, suitable for microbiology, NutriSelect® Plus
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%