Merck
  • Transepithelial electrical resistance (TEER): a functional parameter to monitor the quality of oviduct epithelial cells cultured on filter supports.
Transepithelial electrical resistance (TEER): a functional parameter to monitor the quality of oviduct epithelial cells cultured on filter supports.
Histochemistry and cell biology (2015-08-01)
Shuai Chen, Ralf Einspanier, Jennifer Schoen
ABSTRACT
Cultivation of oviduct epithelial cells on porous filters fosters in vivo-like morphology and functionality. However, due to the optical properties of the filter materials and the cells' columnar shape, cell quality is hard to assess via light microscopy. In this study, we aim to evaluate transepithelial electrical resistance (TEER) measurement as a prognostic quality indicator for the cultivation of porcine oviduct epithelial cells (POEC). POEC were maintained in four different types of media for 3 and 6 w to achieve diverse culture qualities, and TEER was measured before processing samples for histology. Culture quality was scored using morphological criteria (presence of cilia, confluence and cell polarity). We furthermore analyzed the correlation between cellular height (as a measure of apical-basal polarization) and TEER in fully differentiated routine cultures (biological variation) and in cultures with altered cellular height due to hormonal stimulation. Fully differentiated cultures possessed a moderate TEER between 500 and 1100 Ω*cm(2). Only 5% of cultures which exhibited TEER values in this defined range had poor quality. Sub-differentiated cultures showed either very low or excessively high TEER. We unveiled a highly significant (P < 0.0001) negative linear correlation between TEER and epithelial height in well-differentiated cultures (both routine and hormone stimulated group). This may point toward the interaction between tight junction assembly and epithelial apical-basal polarization. In conclusion, TEER is a straightforward quality indicator which could be routinely used to monitor the differentiation status of oviduct epithelial cells in vitro.
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%