Merck
  • Increased Circulating Advanced Oxidation Protein Products and High-Sensitive Troponin T in Cirrhotic Patients with Chronic Hepatitis C: A Preliminary Report.
Increased Circulating Advanced Oxidation Protein Products and High-Sensitive Troponin T in Cirrhotic Patients with Chronic Hepatitis C: A Preliminary Report.
BioMed research international (2015-12-15)
Jolanta Zuwala-Jagiello, Eugenia Murawska-Cialowicz, Monika Pazgan-Simon
ABSTRACT
To investigate the relationship between advanced oxidation protein products (AOPPs) and myocardial injury by comparing the selected biomarker for detecting myocardial injury [high-sensitive troponin T (hs-TnT)] in patients with chronic HCV infection. Eighty-eight patients with cirrhosis and 40 healthy control subjects were enrolled in the study. Circulating levels of AOPPs-albumin (the ratio of AOPPs to albumin content), hs-TnT, tumor necrosis factor α (TNF-α), and high-sensitivity C-reactive protein (hs-CRP) were assessed. Compared with healthy controls, the cirrhotic patients with chronic HCV infection had higher levels of AOPPs-albumin, which were associated with increased hs-TnT. When the presence of ascites was considered, the plasma levels of AOPPs-albumin were higher, as well as TNF-α. AOPPs-albumin positively correlated with hs-TnT level in all cirrhotic patients with chronic HCV infection and this correlation was stronger in decompensated cirrhotic patients. In multivariate logistic regression analysis, the independent factors associated with the presence of ascites were high AOPPs-albumin levels and elevated hs-TnT levels. The simultaneous monitoring of plasma AOPPs and hs-TnT can be helpful for the alterations in myocardial function control in cirrhotic patients with chronic HCV infection.
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%