Merck
Bioactive triterpenoids from Atriplex lasiantha.
Journal of Asian natural products research (2015-03-19)
Basharat Ali, Rehana Tabassum, Naheed Riaz, Asma Yaqoob, Tasneem Khatoon, Rasool Bakhsh Tareen, Abdul Jabbar, Faiz-ul-Hassan Nasim, Muhammad Saleem
ABSTRACT
Chromatographic purification of the ethyl acetate soluble fraction from the methanolic extract of Atriplex lasiantha yielded a new triterpenoid, 7β,15α,16β-trihydroxyolean-12-ene-28,30-dioic acid-3-O-β-D-xylopyranoside (1), along with two known triterpenoids, rotundifolioside I (2) and corchorusin B (3). Structures of the compounds 1-3 were elucidated through sophisticated NMR studies and high resolution mass spectrometry. The three isolates (1-3) were evaluated for antibacterial, antioxidant, and antiurease activities. Compound 2 exhibited the best antibacterial activity against Escherichiacoli with IC50 value of 66.25 μg/ml, whereas, all the tested compounds exhibited antioxidant (IC50 values of 68.7-75.4 μg/ml) and antiurease (IC50 values of 25.5-49.3 μg/ml) activities, respectively.
Sigma-Aldrich
Cerium(IV) sulfate