Merck
  • Fibronectin chorused cohesion between endothelial progenitor cells and mesenchymal stem cells of mouse bone marrow.
Fibronectin chorused cohesion between endothelial progenitor cells and mesenchymal stem cells of mouse bone marrow.
Cellular and molecular biology (Noisy-le-Grand, France) (2015-05-31)
H W Zhang, X L Chen, Z Y Lin, J Xia, J X Hou, D Zhou, Y Xi, M Zhang, J Guo, W Feng, X Y Peng, X W Wu
ABSTRACT
Endothelial progenitor cells (EPCs) could function as niche cells to promote self—renewal of mesenchymal stem cells (MSCs) in the mouse bone marrow. Cohesion was the basis of the two cells to display their biological functions to each other. In this study, we investigated the mechanism of cohesion between MSCs and EPCs. And demonstrated that fibronectin (FN) in EPCs activated the integrin α5β1 of MSCs and further mediated cell-cell cohesion. Integrin α5β1 and its FN ligand played critical roles not only in single—cell line adhesion, but also in adhesion between stem and niche cells. This novel finding is important to understand the cross—talk between MSCs and their niche cells.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fn1
Sigma-Aldrich
MISSION® esiRNA, targeting human FN1
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture